Learn More
BRANDTECH™ BRAND™ Replacement Tip Cones and O-Rings for Transferpette™ S, Transferpette™ Electronic, and Transferpette™ Pro Multichannel Pipettes
Shop All BrandTech Scientific ProductsDescription
- 705677: Includes two replacement tip cones for 0.5 to 10 μL nominal capacity Transferpette™ S, Transferpette™ electronic, and Transferpette™ pro pipettes with dual O-ring design, complete with mounted O-rings
- 705678: Includes two replacement tip cones for 1 to 20 μL nominal capacity Transferpette™ electronic pipettes with dual O-ring design, complete with mounted O-rings
- 705680: Includes two replacement tip cones compatible with older models of BRAND Transferpette™ electronic 1 to 20 μL multichannel pipettes with a single O-ring design
- 703380: Includes 24 O-rings; designed for use with Transferpette™ S, Transferpette™ electronic, and Transferpette™ pro multichannel pipettes with nominal capacities of 0.5 to 10 μL and 1 to 20 μL
Specifications
Specifications
| Description | Replacement tip cones for BRAND Transferpette S and electronic -8/12 multichannel pipettes, 20uL nominal capacity, 1-ring design |
| For Use With | Transferpette Electronic |
| Material | Polypropylene |
| Model | Transferpette -8/12 |
| Sterility | Non-sterile |
| Type | O-ring Kit |
| Volume (Metric) | 20 μL |
| For Use With (Equipment) | Electronic Pipette, 20 μL |
| Includes | Nose cones, O-rings, seals, instructions |
| Quantity | 2 Pack |