Description
- Includes 4 different NTU Standards
- Each standard is supplied in a 60 mL HDPE Bottle
Specifications
Specifications
| Type | Turbidity Standard |
| For Use With (Equipment) | Part Numbers: 3563510 and 3563515 |
| Includes | 4 NTU Standards (20, 100, 400, and 800) in HDPE Bottles |
| For Use With (Application) | Calibration of Oakton T-100 Turbidity Meters |
| Certifications/Compliance | None |
| Volume (Metric) | 60 mL of Each Standard |