Learn More
ProteinSimple RePlex Module for Jess
Supplier:  ProteinSimple RP001
Encompass
This RePlex Module provides all the reagents needed to remove primary and secondary antibodies in a RePlex assay with two immunoassays or immunoassay with Total Protein assay in a single run. Please note this module can only be used with Jess.
Catalog No. NC1839594