Learn More
Sartorius™ ReproEasy Performance Test Pads
GREENER_CHOICE
Catalog No. 14560027 Shop All Sartorius Products
Change view
Click to view available options
Type:
Moisture Analyzer Test Pads, 10/Pk.
Catalog No. Type
14-560-027 Moisture Analyzer Test Pads, 10/Pk.
1 options
Catalog No. 14-560-027 Supplier Sartorius™ Supplier No. YHP02MA
Add to Cart
Add to Cart
Test pads for the periodic review of the performance of MA37/160 moisture analyzer.
  • Allows you to quickly confirm whether a device is working properly and delivers correct results.
  • Use together with the performance test function in moisture analyzer.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) MA37/160 Moisture Analyzer
Quantity 20/Pk.
Type Moisture Analyzer Test Pads, 10/Pk.
WARNING: Cancer - www.P65Warnings.ca.gov