Learn More
Aat Bioquest Resazurin, sodium salt *CAS 62758-13-8*
SDP
Supplier:  Aat Bioquest 15700
Encompass_Preferred
Resazurin is a non-toxic, water-soluble and redox-sensitive dye that changes from its blue/non-fluorescent state to pink/highly-fluorescent upon reduction to resorufin by many biological processes.
Catalog No. 50-195-3577
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.