Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Research™ 3 Neo Pipette Bundle PROMO

Catalog No. 14557321PM Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
1 to 20 μL
10 to 200 μL
50 to 1000 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-557-321PM 50 to 1000 μL
14-557-256PM 1 to 20 μL
14-557-293PM 10 to 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-557-321PM Supplier Eppendorf™ Supplier No. 2231001202

missing translation for 'validMainframeMsg'

Limited Time Promo! Celebrate the Launch of the Eppendorf Research™ 3 neo and receive a FREE ColorTag labeling ring sample pack + ep Dualfilter T.I.P.S.™. Smarter pipetting, brighter workflows.

Research™ 3 Neo Pipette Bundle PROMOs

  • Save up to 50% from January 1 to June 30, 2026!

Includes:

14557256PM:

  • Eppendorf Research™ 3 neo 1-channel 1 to 20 μL, yellow
  • ep Dualfilter T.I.P.S.™, 2 to 20 μL, 53 mm, yellow, colorless tips, PCR clean and sterile (960 tips)
  • ColorTag pipette marking rings sample bag with each size in 2 colors (all 12 colors included)

14557293PM:

  • Eppendorf Research™ 3 neo 1-channel; 10 to 200 μL, yellow
  • ep Dualfilter T.I.P.S.™, 2 to 200 μL, 55 mm, yellow, colorless tips, PCR clean and sterile (960 tips)
  • ColorTag pipette marking rings sample bag with each size in 2 colors (all 12 colors included)

14557321PM:

  • Eppendorf Research™ 3 neo 1-channel; 50 to 1000 μL, blue
  • ep Dualfilter T.I.P.S.™, 50 to 1000 μL, 76 mm, yellow, colorless tips, PCR clean and sterile (960 tips)
  • ColorTag pipette marking rings sample bag with each size in 2 colors (all 12 colors included)

Features:

  • Mechanical air-cushion pipette with 2-button operation (one operating element for volume adjustment/pipetting and one for tip ejection), and customizable settings for easy, ergonomic, and reliable pipetting
  • Spring-loaded tip cones reduce tip attachment and ejection forces
  • Temporary adjustment to increase accuracy for various liquid types, tip geometries, or altitudes without permanently altering the factory calibration
  • Full autoclavability, high UV and chemical resistance
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 8655, ACT (US) Label
Compatible Tips epT.I.P.S.™
Autoclavable Autoclavable
Color Blue
Disposable Reusable
For Use With Pipette Tips
Grip Type Ergonomic
Material Polypropylene
Number of Channels 1
Product Line Research
Programmability No
Sterility Non-sterile
Type Adjustable Single Channel Pipette
Volume (Metric) 50 to 1000 μL
Finger Style Ergonomic
For Use With (Equipment) Pipette Tip 1000 μL
Includes ep Dualfilter T.I.P.S.™, 50 to 1000 μL, 76 mm, Yellow, Colorless Tips, PCR Clean and Sterile (960 Tips); ColorTag Pipette Marking Rings Sample Bag with Each Size in 2 Colors (All 12 Colors Included)
Quantity 1 Pk.
Warranty 24 Months
Length (Metric) 71 mm
Weight (Metric) 80 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.