Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Research™ Plus Multichannel Multi-Pack PROMO

Catalog No. 13690050PM Shop All Eppendorf North America Products
Change view
Click to view available options
Number of Channels:
8
12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Number of Channels
13-690-050PM 12
13-690-047PM 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-690-050PM Supplier Eppendorf™ Supplier No. 2231001177

missing translation for 'validMainframeMsg'

Boost 96-well throughput with Research™ Plus multichannel bundles: get 3 ultralight, fully autoclavable pipettes (10, 100, 300 μL) in 8- or 12-channel formats. Spring-loaded tip cones and PhysioCare™ ergonomics deliver fast, fatigue-free precision.

Eppendorf Research™ Plus 8-Channel (10 μL, 100 μL, 300 μL) Pipettes Promotion Includes:

  • 3 x Research™ Plus 8-channel pipettes (10 μL, 100 μL, 300 μL)

Eppendorf Research™ Plus 12-Channel (10 μL, 100 μL, 300 μL) Pipettes Promotion Includes:

  • 3 x Research™ Plus 12-channel pipettes (10 μL, 100 μL, 300 μL)

Eppendorf Research™ Plus Pipette Features:

  • Volume adjustment feature allows desired volume to be reached in few quick turns
  • Control button and ejector's low operating force are positioned for ergonomics
  • 4-digit, magnifying-shape volume display
  • Calibration display and adjustment opening for specific liquids and volumes
  • Uses renowned PhysioCare Concept™ thus reduces strain on hand and arm during pipetting, maintains balance, minimal user exertion
  • Quick lock makes lower part particularly easy to remove and service (not available for 5 and 10 mL pipettes)
  • Spring loaded tip cone promotes tight fit to tip (not available for 5 and 10 mL pipettes)
  • Has ability to be autoclaved without disassembling
TRUSTED_SUSTAINABILITY

Specifications

Accuracy ±8% at 0.5 μL, ±2.5% at 1 μL, ±1.5% at 5 μL, ±1% at 10 μL
Autoclavable Autoclavable
Color Medium Gray
Number of Channels 12
Type Mechanical Pipette
Volume (Metric) 10, 100, 300 μL
Display 4 Digits
Finger Style Ergonomic
Increments 0.5, 1, 5, 10 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.