Learn More
Eppendorf™ Research™ Plus Single-Channel Multi-Pack PROMO
Catalog No. 05403151PM1 Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
10, 100, 1000 μL
10, 20, 200, 1000 μL
2.5, 10, 100, 1000 μL
2.5, 20, 200, and 1000 μL
20, 200, 1000 μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
05-403-151PM1 2.5, 10, 100, 1000 μL
31-160-00015P 2.5, 20, 200, and 1000 μL
14-285-904PM 10, 20, 200, 1000 μL
13-684-250PM 10, 100, 1000 μL
13-684-251PM1 20, 200, 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 05-403-151PM1 Supplier Eppendorf™ Supplier No. 2231001167
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Essential bundles! Start, Expand, or Replace your pipettes with deals on 3- or 4-pack sets covering 2.5 to 1000 μL. Ultralight, adjustable volume, fully autoclavable bodies and spring-loaded tip cones deliver fatigue-free precision at every volume.
4 Pack Deals:
Eppendorf Research™ Plus 4-Pack and Carousel Stand Bundle (Option 1) Includes:
  • 4 x Research™ Plus pipettes (2.5/10/100/1000 μL) with carousel stand
Eppendorf Research™ Plus 4-Pack and Carousel Stand Bundle (Option 2) Includes:
  • 4 x Research™ Plus pipettes (2.5/20/200/1000 μL) with carousel stand
Eppendorf Research™ Plus 4-Pack and Carousel Stand Bundle (Option 3) Includes:
  • 4 x Research™ Plus pipettes (10/20/200/1000 μL) with carousel stand
3 Pack Deals:
Eppendorf Research™ Plus 3-Pack Bundle (Option 1) Includes:
  • 3 x Research™ Plus pipettes (10/100/1000 μL)
Eppendorf Research™ Plus 3-Pack Bundle (Option 2) Includes:
  • 3 x Research™ Plus pipettes (20/200/1000 μL)
Eppendorf Research™ Plus Features:
  • Volume adjustment feature allows desired volume to be reached in few quick turns
  • Control button and ejectors low operating force are positioned for ergonomics
  • 4-digit, magnifying-shape volume display
  • Calibration display and adjustment opening for specific liquids and volumes
  • Uses renowned PhysioCare Concept™ thus reduces strain on hand and arm during pipetting, maintains balance, minimal user exertion
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Number of Channels 1
Product Line Research Plus
Type Pipette Package
Volume (Metric) 2.5, 10, 100, 1000 μL