Learn More
Eppendorf™ Research™ Plus Single-Channel Multi-Pack PROMO
Catalog No. 3116000015P Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
10, 100, 1000 μL
10, 20, 200, 1000 μL
2.5, 10, 100, 1000 μL
2.5, 20, 200, and 1000 μL
20, 200, 1000 μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
31-160-00015P 2.5, 20, 200, and 1000 μL
05-403-151PM1 2.5, 10, 100, 1000 μL
14-285-904PM 10, 20, 200, 1000 μL
13-684-250PM 10, 100, 1000 μL
13-684-251PM1 20, 200, 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 31-160-00015P Supplier Eppendorf™ Supplier No. 2231001168
Essential bundles! Start, Expand, or Replace your pipettes with deals on 3- or 4-pack sets covering 2.5 to 1000 μL. Ultralight, adjustable volume, fully autoclavable bodies and spring-loaded tip cones deliver fatigue-free precision at every volume.
4 Pack Deals:
Eppendorf Research™ Plus 4-Pack and Carousel Stand Bundle (Option 1) Includes:
  • 4 x Research™ Plus pipettes (2.5/10/100/1000 μL) with carousel stand
Eppendorf Research™ Plus 4-Pack and Carousel Stand Bundle (Option 2) Includes:
  • 4 x Research™ Plus pipettes (2.5/20/200/1000 μL) with carousel stand
Eppendorf Research™ Plus 4-Pack and Carousel Stand Bundle (Option 3) Includes:
  • 4 x Research™ Plus pipettes (10/20/200/1000 μL) with carousel stand
3 Pack Deals:
Eppendorf Research™ Plus 3-Pack Bundle (Option 1) Includes:
  • 3 x Research™ Plus pipettes (10/100/1000 μL)
Eppendorf Research™ Plus 3-Pack Bundle (Option 2) Includes:
  • 3 x Research™ Plus pipettes (20/200/1000 μL)
Eppendorf Research™ Plus Features:
  • Volume adjustment feature allows desired volume to be reached in few quick turns
  • Control button and ejectors low operating force are positioned for ergonomics
  • 4-digit, magnifying-shape volume display
  • Calibration display and adjustment opening for specific liquids and volumes
  • Uses renowned PhysioCare Concept™ thus reduces strain on hand and arm during pipetting, maintains balance, minimal user exertion
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Number of Channels 1
Product Line Research Plus
Type Pipette Package
Volume (Metric) 2.5, 20, 200, and 1000 μL