Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Residual Solvents Class 2 - Mix A, Restek
SDP

Catalog No. 06729402 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-729-402 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-729-402 Supplier Restek Corporation Supplier No. 36271
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

Analyte or Component Names 15 components:
acetonitrile (2.05mg/mL)
chlorobenzene (1.80mg/mL)
cyclohexane (19.40mg/mL)
cis-1,2-dichloroethene (4.70mg/mL)
trans-1,2-dichloroethene (4.70mg/mL)
1
Physical Form Liquid
Element Acetonitrile, Chlorobenzene, Cyclohexane, cis-1,2-dichloroethene, trans-1,2-dichloroethene
Solvent or Matrix Dimethyl Sulfoxide
Quantity 1 mL
Recommended Storage Store in a cool dry ventilated location. Keep containers closed away from heat, sparks, and flame.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.