Learn More
Bio Rad Laboratories RESIN BIO-REX 70
Supplier:  Bio Rad Laboratories 1435852
Encompass
Media, Biotechnology grade; Bio-Rad; 70 resin; 100g; Sodium; 200-400 dry mesh; 45-75um wet bead
Catalog No. 50-901-2936