Learn More
Selleck Chemical LLC Resiquimod 50mg 144875-48-9 R-848
SDP
Supplier:  Selleck Chemical LLC S813350MG
Encompass_Preferred
Resiquimod (R-848) is an immune response modifier that acts as a potent TLR 7/TLR 8 agonist that induces the upregulation of cytokines such as TNF-?, IL-6 and IFN-?. Resiquimod reduces hepatitis C virus (HCV) infection. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5693
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.