Learn More
Vernier™ Resistivity Rod Set
SDPSCI_ED
Catalog No. S15997ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Current Probe, Instrumentation Amplifier
Catalog No. For Use With (Equipment)
S15997ND Current Probe, Instrumentation Amplifier
1 options
Catalog No. S15997ND Supplier Vernier™ Supplier No. RRS
Add to Cart
Add to Cart
Includes
Resistivity Rod Set
Use this set of rods with the Instrumentation Amplifier and the Current Probe to investigate electrical resistance as a function of conductor length, diameter, and material. This lab is featured in our lab manual.
  • Inlcudes 4 brass rods: 2.38 mm (3/32 in.) diameter, 3.18 mm (1/8 in.) diameter, 3.97 mm (5/32 in.) diameter, 4.76 mm (3/16 in.) diameter
  • One stainless steel rod, one music wire rod, one aluminum rod, one copper rod all with 3.18 mm (1/8 in.) diameter
Warranty and Services
5-year limited warranty
Specifications
Includes 4 brass rods, 1 stainless steel rod, 1 music wire rod, 1 aluminum rod, 1 copper rod
Material Brass, Stainless Steel, Aluminum, and Copper
Warranty 5 Year Limited
For Use With (Application) Educational use only
For Use With (Equipment) Current Probe, Instrumentation Amplifier
Length (Metric) 46 cm
WARNING: Cancer - www.P65Warnings.ca.gov