Learn More
Cayman Chemical Resolvin D1, 872993-05-0, 10 g
SDP
Supplier:  Cayman Chemical 1001255410
Encompass_Preferred
Molecular Formula C22H32O5, Purity ≥95%, Formula Weight 376.5, Synonyms: 17(S)-Resolvin D1, RvD1; A potent anti-inflammatory mediator derived physiologically from the sequential oxygenation of DHA by 15- and 5-LO; reduces human leukocyte transendothelial migration (EC50 = ~30 nM) and limits leukocyte infiltration in a mouse model of peritonitis.
Catalog No. NC0069649
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.