Learn More
United Scientific Supplies Resonance Apparatus
SDPSCI_ED
Catalog No. S82875 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Material:
Acrylic
Catalog No. Material
S82875 Acrylic
1 options
This item is not returnable. View return policy
Prop 65 Warning
Catalog No. S82875 Supplier United Scientific Supplies Supplier No. RSAPP01
Add to Cart
Add to Cart
This item is not returnable. View return policy
Prop 65 Warning
Resonance Apparatus allows students to investigate resonant behavior.
  • The Resonance Apparatus allows students to investigate the resonant behavior of an enclosed air column when it is stimulated by an external sound source.
  • The air column lengths for multiple resonances at various frequencies can be found and the resulting data can be used to determine the speed of sound.
  • A cylindrical acrylic tube is mounted vertically on a stand base and rod.
  • The tube is open at the top and closed at the bottom by a rubber stopper with a hose barb.
  • A scale is printed along the length of the tube.
  • Flexible tubing connects the bottom of the tube to a large reservoir.
  • The reservoir and tube are partially filled with water, creating an air space in the tube above the water level.
  • The length of the space can be varied by adjusting the height of the reservoir on the support rod.
  • A suitable sound source, which can be a tuning fork (not included) or a tone generator (not included), is held close to the mouth of the tube while the length of the air column is varied.
  • Points of resonance where the sound in the tube becomes louder can be heard and measured.
  • The sound level may also be recorded for later analysis using probeware.
Specifications
Product Type Resonance Apparatus
Material Acrylic
WARNING: Reproductive Harm - www.P65Warnings.ca.gov