Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Resorufin Butyrate, Fluorogenic Lipase and Cholinesterase Substrate, 250 mg

Catalog No. 502705195
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5195 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5195 Supplier Abcam Supplier No. AB145435250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorogenic substrate for lipase and cholinesterase that fluoresces upon hydrolysis.

  • Molecular weight: 283.28 Da
  • Purity: >95%
  • Functional category: Fluorogenic lipase and cholinesterase substrate
  • Key properties: Fluoresces on hydrolysis
  • Analog of: Resorufin methyl ether (ab145372)

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at -20°C.
Target Molecule Lipase, Cholinesterase
Buffer 100% Resorufin butyrate
Shipping Condition Ambient
Detection Method Fluorescence
Formulation 100% Resorufin butyrate
Quantity 250 mg
For Use With (Application) Lipase assay, Cholinesterase assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.