Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Resorufin Butyrate, Fluorogenic Lipase Substrate, 25 mg

Catalog No. 502705194
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5194 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5194 Supplier Abcam Supplier No. AB14543525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorogenic substrate for the detection of lipase and cholinesterase activity.

  • Molecular Weight: 283.28 Da
  • Purity: >95%
  • Application: Fluorogenic lipase and cholinesterase substrate; fluoresces on hydrolysis
  • Analog: Resorufin methyl ether (ab145372) analog

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Lipase, Cholinesterase
Substrate Properties Chemical Substrate
Substrate Resorufin butyrate
Content And Storage Store at -20°C.
Shipping Condition Ambient
Buffer 100% Resorufin butyrate
Detection Method Fluorescence
Formulation 100% Resorufin butyrate
Quantity 25 mg
For Use With (Application) Lipase assay, cholinesterase assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.