Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Resorufin Methyl Ether, Fluorometric Cytochrome P450 Substrate, 1 mg
Description
- Molecular Weight: 227.21 Da
- Applications: Fluorometric competitive cytochrome P450 substrate
- Selectivity: CYP1A (Ki = 20 μM, dealkylation 0.5 μM substrate)
- Composition: 100% Methylresorufin
- CAS Number: 5725-89-3
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | Cytochrome P450 |
| Substrate | Cytochrome P450 |
| Buffer | 100% Methylresorufin |
| Shipping Condition | Blue Ice |
| Detection Method | Fluorescence |
| Content And Storage | Store at -20°C. |
| Formulation | 100% Methylresorufin |
| Quantity | 1 mg |
| For Use With (Application) | Cytochrome P450 assay |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?