Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva RESOURCE™ Columns for HIC

Catalog No. p-3676546 Shop All Cytiva Products
Change view
Click to view available options
Bed Volume:
1 mL
1 x 1 mL
Ligand Type:
Isopropyl
Phenyl
Product Type:
RESOURCE ISO column
RESOURCE PHE column
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Volume Ligand Type Product Type
45-000-204 1 mL Phenyl RESOURCE PHE column
45-500-108 1 x 1 mL Isopropyl RESOURCE ISO column
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-000-204 Supplier Cytiva Supplier No. 17118601
Only null left
Add to Cart
Add to Cart

Includes

Connectors for high and low pressure chromatography equipment; Instructions.

Excellent for polishing in a purification process

  • Prepacked columns for purification of proteins and peptides with hydrophobic interaction chromatography (HIC)
  • High performance at low back pressure, even with high sample loads of up to 25mg of protein
  • Ligands coupled to 15μm monodispersed rigid polystyrene/divinylbenzene matrix
  • Designed for low- to medium- pressure systems ranging from peristaltic pump to AKTA design and FPLC Systems
  • RESOURCE PHE will have strongest hydrophobicity followed by RESOURCE ISO and RESOURCE ETH, successively
  • Material of column: Polyetheretherketone (PEEK)
  • Bed dimensions 6.4 x 30mm
  • Maximum pressure 15 bar
  • Flow rate: 0.8 to 4.8mL/min
  • Storage: 4°C to 30°C, 20% Ethanol

Media

  • Matrix: Polystyrene/divinylbenzene
  • Stable in common buffers, chaotropic agents, detergents, and polar organic solvents
  • Average particle size: 15Glc mum
  • pH Stability Working Range: 2 to 12
  • Flow velocity: 150cm/h - 900cm/h

Resource HIC Test Kit (Mfr. No. 17-1187-01) includes one each of 1mL Resource ETH, Resource ISO and Resource PHE columns

TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 1 mL
Diameter (English) Inner 3.937 in.
Diameter (Metric) Inner 10 cm
Product Type RESOURCE PHE column
Flow Rate 150 to 900 cm/hr
Quantity 1 mL
Height (English) 3.937 in.
Height (Metric) 10 cm
Includes Connectors for high- and low-pressure chromatography equipment and instructions
Ligand Type Phenyl
Material PEEK
Max. Pressure 15 bar
Particle Size 15 to 15 μm
pH Range 2 to 12
Support Type Polystyrene/Divinylbenzene
Storage Requirements 4°C to 30°C, 20% Ethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.