Learn More
Dentec Safety™ Respirator Filter Holder for Particulate Pads
Shop All Dentec Safety Specialists Inc. Products
Description
- Will not obstruct field of vision
- Holder has spacers that create a space between the filter material and the holder, allowing the user to access the entire filter material increasing filter life while reducing breathing resistance and increasing productivity.
Specifications
Specifications
| Material | Plastic |
| For Use With (Equipment) | Masks |
| Quantity | 6/Pk. |