Learn More
Essendant RESPIRATORDUAL CARTRGGY
Supplier:  Essendant MMM52P71
Encompass_Preferred
The 3M 5000 Series Half Facepiece Disposable Respirator Assembly with Organic Vapor and P95 Particulate Filters Respiratory Protection (51P71 52P71 53P71) helps provide protection against certain airborne contaminants This respirator is NIOSH approved for respiratory protection against certain organic vapors and particulates up to ten times the permissible exposure limit (PEL) Half Facepiece respirator may be used in a variety of workplace applications including spray painting pesticides agriculture construction and transportation This disposable respirator assembly offers greater convenience by reducing the need for long-term maintenance and is an ideal choice for intermittent use NIOSH color code of Black Respirator Type- Half-Face Mask Rating- P95 Airborne Contaminant Resistance- Organic Vapors Material(s)- Thermoplastic Elastomer (TPE)
Catalog No. 50-000-35809
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.