Learn More
Restek™ Resprep-C18 and Resprep-C8 SPE Discs
SDP
Catalog No. 06611915 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
12 Pk.
Catalog No. Quantity
06-611-915 12 Pk.
1 options
Catalog No. 06-611-915 Supplier Restek™ Supplier No. 25988
Glass fiber disks embedded with C18 or C8 bonded silica
  • Extract semivolatile organic compounds
  • Deep-pore design reduces clogging and allows faster flow rates
  • Meets requirements for EPA methods 525.1, 506, 550.1 and 549.1
  • Lower cost than PTFE disks
Specifications
Type Resprep SPE Disks
Quantity 12 Pk.
Diameter (Metric) Outer 90 mm