Description
- Cap material: Polypropylene
- Preassembled (short cap and liner)
Specifications
Specifications
| Material (Liner) | PTFE/Silicone |
| Color | Yellow |
| Material | Polypropylene |
| Pre-slit | Yes |
| Septum | PTFE/Silicone/PTFE |
| Quantity | 1000/Pk. |
| Closure Size | 9 mm |