Learn More
Restek™ ASI Check Valves for Agilent 1050 and 1100
SDP
For Agilent 1050, 1100 HPLC Systems.
Specifications
For Use With (Equipment) Agilent 1050 and 1100 HPLC System
View More Specs
Products 4
Catalog Number Mfr. No. Type Material Price Quantity & Availability  
Catalog Number Mfr. No. Type Material Price Quantity & Availability  
06-712-727
SDP
View Documents
Restek™
25171
Outlet Cartridge Sapphire and Ruby Ball and Seat Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
06-712-729
SDP
View Documents
Restek™
25173
Outlet Check Valve (Cartridge and Housing) Ceramic Ball and Seat Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
06-712-730
SDP
View Documents
Restek™
25174
Outlet Cartridge Ceramic Ball and Seat Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
06-712-728
SDP
View Documents
Restek™
25172
Outlet Housing N/A Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description
Description
  • Valves do not leak even at pressures up to 12,000psi (82,737 kPa)
  • Valves close properly even without a high backpressure force
  • Cartridge design allows easy, convenient replacement of worn or damaged valves
Specifications
Specifications
Agilent 1050 and 1100 HPLC System
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.