Learn More
Restek™ ASI Check Valves for Waters Alliance Model 2690
SDP
Catalog No. 06712718 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Material:
Ceramic Ball and Seat
Sapphire and Ruby Ball and Seat
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
06-712-718 Ceramic Ball and Seat
06-712-716 Sapphire and Ruby Ball and Seat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-712-718 Supplier Restek™ Supplier No. 25162
For Waters Alliance Model 2690 and 2695.
  • Valves do not leak even at pressures up to 12,000psi (82,737 kPa)
  • Valves close properly even without a high backpressure force
  • Cartridge design allows easy, convenient replacement of worn or damaged valves
Specifications
For Use With (Equipment) Waters Alliance Model 2690 and 2695
Material Ceramic Ball and Seat
Type Inlet and Outlet Cartridge