Learn More
Restek™ ASI Piston Seals
SDP
For Agilent HPLC Systems.
$112.00 - $1095.00
Specifications
For Use With Agilent 1050 and 1100 HPLC system
Material PTFE
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
06-712-731
SDP
View Documents
Restek™
25175
ASI Piston Seal
Each for $112.00
Only null left
Add to Cart
06-712-732
SDP
View Documents
Restek™
25176
Piston Seal
Each for $1,095.00
Only null left
Add to Cart
Description
Description
  • Constructed of PTFE material with an additive that reduces seal's tendency to “cold flow” and improves their wear characteristics
  • More resistant to any HPLC solvent than UHMWPE seals
Specifications
Specifications
Agilent 1050 and 1100 HPLC system
PTFE
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.