Learn More
Restek™ Back Pressure Regulators
SDP
Improves detector performance by preventing bubble formation in the detector flow cell.
Supplier:  Restek™ 25018
Catalog No. 06-712-599
Description
Description
  • Useful in post-column reaction lines and between detectors and fraction collectors in preparatory work
  • Backpressure regulators are adjustable to ensure constant backpressure over a wide range of mobile phase viscosities and flow rates
  • End-of-line model is available with 1/4-28 plastic flange-type fittings or high-pressure 0.0625 in. compression fittings and adjusts from 15 to 60psi (103 to 414 kPa)
  • Flow-through design has 0.0625 in. compression fittings and is adjustable from 7 to 75psi (48 to 517 kPa)
Specifications
Specifications
Back Pressure Regulator
15 to 60 psi
0.25 to 28 plastic flange-type fittings or high-pressure 0.0625 in. compression fitting
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.