Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Canister Carrying Case
SDP

Catalog No. 06712239 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
Canister Carrying Case
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
06-712-239 Canister Carrying Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-712-239 Supplier Restek™ Supplier No. 24226
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heavy-duty, all-aluminum design fits two 6 L SilcoCan or TO-Can canisters tightly without foam.

  • No organic contaminants from foam or plastics
  • Inside Dimensions (L x W x H): 46 x 23 x 32cm
  • Weight: 4kg (9 lb.)

Specifications

For Use With (Equipment) Two 6L SilcoCan™ or TO-Can™ canisters tightly without foam
Weight (English) 9 lb.
Weight (Metric) 4.08 kg
Type Canister Carrying Case
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.