Learn More
Restek™ Convenience Kits with Vial, Cap, and Septa: Untreated
SDP
Catalog No. 06718475 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Color:
Amber
Clear
Quantity:
100/Pk.
1000/Pk.
Septum:
Rubber
Silicone
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity Septum
06-718-475 Clear 100/Pk. Rubber
06-718-476 Clear 1000/Pk. Rubber
06-718-477 Amber 100/Pk. Rubber
06-718-478 Amber 1000/Pk. Rubber
06-718-980 Clear 1000/Pk. Silicone
06-718-982 Amber 1000/Pk. Silicone
06-718-979 Clear 100/Pk. Silicone
06-718-981 Amber 100/Pk. Silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 06-718-475 Supplier Restek™ Supplier No. 21196
Add to Cart
Add to Cart
Available with or without white graduated marking spots
  • Vials packaged in a clear-lid tray
  • Caps with septa packaged in a plastic bag
Specifications
Color Clear
Volume (Metric) 2 mL
Septum Rubber
Material (Seal) Silver
Closure Material PTFE
Closure Type Crimp Cap
Quantity 100/Pk.
Closure Size 11 mm