Learn More
Restek™ Diskcover-47 Extraction Disk Holder
SDP
Catalog No. 06712149 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
06-712-149 Each
1 options
Catalog No. 06-712-149 Supplier Restek™ Supplier No. 24020
Compatible with most vacuum manifold systems that accept 0.125 in. male luer fittings.
  • Sample can be automatically introduced via 0.125 in. PTFE tubing or from the optional Diskcover-47 reservoir
Specifications
For Use With (Equipment) Most Vacuum Manifold Systems That Accept 0.12 in. Male Luer Fittings
Type Extraction Disk Holder
Quantity Each