Description
- Certified reference material (CRM) manufactured and QC-tested in Restek's ISO-accredited labs
- In acetonitrile, 1mL/ampule
Specifications
Specifications
| Quantity | 1 mL/Ampule |
| Analyte or Component Names | 4-Androstene-3,17-dione (63-05-8), 100 μg/mL; Equilin (474-86-2), 200 μg/mL; 17-beta-Estradiol (50-28-2), 250 μg/mL; Estriol (50-27-1), 200 μg/mL; Estrone (53-16-7), 200 μg/mL; 17-alpha-Ethynylestradiol (57-63-6), 350 μg/mL; Testosterone (58-22-0), 100 μg/mL |
| Type | EPA 539 Calibration Stock Standard |