Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ High-capacity and ECD Vent Traps
SDP

Catalog No. 06717795 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
ECD Vent Trap
High Capacity Split Vent Trap
Packaging Quantity:
1 Each
5/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Packaging Quantity
06-717-795 High Capacity Split Vent Trap 5/Pk.
06-710-915 High Capacity Split Vent Trap 1 Each
06-711-822 ECD Vent Trap 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-717-795 Supplier Restek™ Supplier No. 20699
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes connecting lines and mounting kit.

  • High-Capacity Split Vent Trap reduces the release of hazardous materials from the capillary split vent into the lab
  • ECD Vent Trap reduces the release of hazardous materials from the ECD vent into the lab
  • Fittings: 1/8 in.
  • Dimensions: 15.2 x 2.5cm (6 x 1 in.)

Specifications

Connection Type 0.12 in. Fitting
Type High Capacity Split Vent Trap
Includes Connecting lines and mounting kit
Packaging Quantity 5/Pk.
Length (English) 6 in.
Length (Metric) 15.2 cm
Diameter (English) Outer 1 in.
Diameter (Metric) Outer 2.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.