Learn More
Description
Used with the Restek Opti-Cap Bottle Top (06712843)
Specifications
Specifications
| Thread Size (Metric) | 65 mm |
| Neck Finish | 65 mm |
| For Use With (Equipment) | 4L Solvent bottle |
| Product Type | Hub Cap Adapter |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
