Learn More
Restek Corporation Restek™ Hydroguard™ Fused Silica Transfer Tubing
SDP
Catalog No. 06708193 Shop All Restek Corporation Products
Encompass_Preferred
Tubing; Hydroguard Fused Silica; Restek; Deactivated high-density surface resists hydrolysis; For transfer lines, purge and trap; 60m length banded in 5m sections; 0.53mm I.D.; 0.69mm O.D. (plus or minus 0.05mm)
Catalog No. 06-708-193 Supplier Restek Corporation Supplier No. 10090
Tubing; Hydroguard Fused Silica; Restek; Deactivated high-density surface resists hydrolysis; For transfer lines, purge and trap; 60m length banded in 5m sections; 0.53mm I.D.; 0.69mm O.D. (plus or minus 0.05mm)