Learn More
Restek Corporation Restek™ Hydroguard MXT Guard Columns and Transfer Lines
SDP
Supplier:  Restek Corporation 70081600
Encompass_Preferred
Column, retention gap; Restek; MXT; Passivated stainless steel; Wide range of applications; Use with most common solvents; Max. temp.: 430deg.C; Nominal ID: 0.53mm; Nominal OD: 0.74 plus/minus 0.025mm; Length: 5m; Pack of 6
Catalog No. 06-713-277
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.