Learn More
Restek Corporation Restek™ Intermediate-Polarity Deactivated MXT Guard Columns and Transfer Lines
SDP
Catalog No. 06713273 Shop All Restek Corporation Products
Encompass_Preferred
Column, retention gap; Restek; MXT; Passivated stainless steel; Wide range of applications; Use with most common solvents; Max. temp.: 430deg.C; Nominal ID: 0.53mm; Nominal OD: 0.74 plus/minus 0.025mm; Length: 10m; Pack of 6
Catalog No. 06-713-273 Supplier Restek Corporation Supplier No. 70047600
Column, retention gap; Restek; MXT; Passivated stainless steel; Wide range of applications; Use with most common solvents; Max. temp.: 430deg.C; Nominal ID: 0.53mm; Nominal OD: 0.74 plus/minus 0.025mm; Length: 10m; Pack of 6