Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ MicroPulse Pulse Dampers
SDP

Compact unit can be placed almost anywhere

$889.00 - $1015.00

Specifications

Type Minipulse Pulse Dampener
For Use With (Application) For Applications Where Minimizing The Total System Volume is Critical
Volume (Metric) 160 μL
View More Specs

Products 2
Catalog Number Mfr. No. Material Pressure Price Quantity & Availability  
Catalog Number Mfr. No. Material Pressure Price Quantity & Availability  
06-712-785
SDP
View Documents
Restek™
25238
Stainless Steel 6000 psi
Each for $889.00
Only null left
Edge
Add to Cart
 
06-712-786
SDP
View Documents
Restek™
25239
PEEK 5000 psi
Each for $1,015.00
Only null left
Add to Cart
 
Description

Description

The MiniPulse pulse damper is ideal for applications where minimizing the total system volume is critical.

  • Compact unit
  • Small
  • 160μL dead volume at atmospheric pressure
  • Diameter: 2.5 x 1.5 in.

Specifications

Specifications

Minipulse Pulse Dampener
For Applications Where Minimizing The Total System Volume is Critical
160 μL
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.