Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Graduated Safety-Coated Bottles
SDP

Graduated, safety-coated bottles with GL-45 threads.

$144.10 - $212.00

Specifications

Type Graduated Bottle
For Use With (Application) Mobile phase management
Includes Solid cap
Material Glass
Thread Size GL-45
View More Specs

Products 2
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
06-712-847
SDP
View Documents
Restek™
25304
1 L
Each for $144.10
Only null left
Add to Cart
 
06-712-848
SDP
View Documents
Restek™
25305
2 L
Each for $212.00
Only null left
Add to Cart
 
Description

Description

Available as 1 L or 2 L bottles.
Specifications

Specifications

Graduated Bottle
Solid cap
GL-45
Mobile phase management
Glass
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.