Learn More
Restek™ Pesticide Residue Cleanup SPE Cartridges
SDP
Catalog No. 06729305 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Sorbent:
CarboPrep PSA
Catalog No. Sorbent
06-729-305 CarboPrep PSA
1 options
Catalog No. 06-729-305 Supplier Restek™ Supplier No. 26128
For use in multiresidue pesticide analysis to remove matrix interferences.
  • Convenient, multiple adsorbent beds in a single cartridge
  • Ideal for cleanup of dietary supplement extracts
Specifications
For Use With (Application) For Use in Multiresidue Pesticide Analysis to Remove Matrix Interferences
Sorbent CarboPrep PSA
Type SPE Cartridge