Learn More
Restek™ Photoionization Lamps
SDP
Catalog No. 06712127 Shop All Restek Corporation Products
Encompass
Change view
Click to view available options
Product Type:
Photoionization Lamp
Catalog No. Product Type
06-712-127 Photoionization Lamp
1 options
Catalog No. 06-712-127 Supplier Restek™ Supplier No. 23020
Higher output makes it ideal for detection of BTEX compounds.
UV energy emitting PID lamps for use with ELCD detectors.
Specifications
For Use With (Equipment) HNU and SRI detectors
Output 10/10.6 eV
Product Type Photoionization Lamp