Learn More
Restek™ Polishing Kit for Photoionization Lamps
SDP
Catalog No. 06710895 Shop All Restek Corporation Products
Encompass
Change view
Click to view available options
Product Type:
PID Lamp Polishing Kit
Catalog No. Product Type
06-710-895 PID Lamp Polishing Kit
1 options
Catalog No. 06-710-895 Supplier Restek™ Supplier No. 20674
Used to clean the outside of the PID lamp window to restore the output.
Includes iron oxide cleaning compound, swabs and instruction.
Specifications
For Use With (Equipment) Photoionization lamp
Includes Iron Oxide cleaning compound, swabs and instructions
Product Type PID Lamp Polishing Kit