Learn More
Restek™ Pressure Gauge Kit for Thermal Gas Purifiers
SDP
Catalog No. 06711712 Shop All Restek Corporation Products
Change view
Click to view available options
Type:
Gas Pressure Gauge Kit
Catalog No. Type
06-711-712 Gas Pressure Gauge Kit
1 options
Catalog No. 06-711-712 Supplier Restek™ Supplier No. 21657
Only null left
Add to Cart
Add to Cart
Use an in-line pressure gauge to indicate when thermal gas purifier getter tube should be replaced
  • Includes 1/8 in. tee and 0-100 psi (0-689k Pa) gauge
Specifications
Type Gas Pressure Gauge Kit
For Use With (Application) Use an in-line pressure gauge to indicate when a thermal gas purifier getter tube should be replace
For Use With (Equipment) Thermal gas purifier
Includes 0.12 in. Tee and 0 to 100psi gauge