Learn More
Restek™ Q-sep™ QuEChERS dSPE Tubes for Extract Clean-Up
SDP
Fast, simple sample prep for multiresidue pesticide analysis.
$366.00 - $471.50
Specifications
For Use With (Application) Fast, Simple Sample Prep for Multi Residue Pesticide Analysis
Type Q-sep QuEChERS dSPE Micro-centrifuge Tube
Method AOAC 2007.01
View More Specs
Products 4
Catalog Number Mfr. No. Material Shake Time Sorbent Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Material Shake Time Sorbent Volume (Metric) Price Quantity & Availability  
06-729-300
SDP
View Documents
Restek™
26123
150mg MgSO4 2 min. Ultra pure, PSA, GCB 2 mL
Pack of 100 for $403.00
Only null left
Edge
Add to Cart
06-729-301
SDP
View Documents
Restek™
26124
150mg MgSO4 0.5 min. Ultra pure, PSA 2 mL
Pack of 100 for $366.00
Only null left
Add to Cart
06-729-302
SDP
View Documents
Restek™
26125
150mg MgSO4 0.5 min. Ultra pure, PSA, C18 2 mL
Pack of 50 for $366.00
Only null left
Edge
Add to cart
06-729-303
SDP
View Documents
Restek™
26126
900mg MgSO4 2 min. Ultra pure, PSA, GCB 15 mL
Pack of 50 for $471.50
Only null left
Edge
Add to cart
Description
Description
QuEChERS methods are fast, easy, and cost-effective, and Restek Q-sep products make QuEChERS procedures even simpler. All extraction salts, sorbents, and sample tubes are included. Multiple sorbents are used to extract different types of interferences.
  • Ready-to-use tubes, no glassware required
  • Preweighed, ultra-pure sorbents
  • Packaged in foil subpacks of 10 for enhanced protection and storage stability
  • Support original unbuffered, AOAC (2007.01), European (EN 15662), and mini-multiresidue QuEChERS methods
Specifications
Specifications
Fast, Simple Sample Prep for Multi Residue Pesticide Analysis
Q-sep QuEChERS dSPE Micro-centrifuge Tube
AOAC 2007.01
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.