Learn More
Restek™ Refillable Hydrocarbon Traps
SDP
Catalog No. 06710858 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
Carbon Refill
Hydrocarbon Trap
Includes:
0.125 in. Fitting
0.25 in. Fitting
Two recharges
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Includes
06-710-858 Carbon Refill Two recharges
06-711-817 Hydrocarbon Trap 0.125 in. Fitting
06-711-818 Hydrocarbon Trap 0.25 in. Fitting
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-710-858 Supplier Restek™ Supplier No. 20626
Add to Cart
Add to Cart
Ideal for use with purge and trap systems to stop carrier gas interferences
  • Remove trace organic impurities from carrier gas to 0.1ppm where input does not exceed 100ppm
  • A 20μm frit prevents system contamination from purifier particles
  • Rechargeable and refillable
  • 1 pt. Carbon Refill with two recharges sold separately
  • Choice of 1/8 or 1/4 in. fittings
Order Info
Dimensions: 9 1/4L x 2 1/4 in. dia. (23.5 x 5.1cm).
Specifications
Type Carbon Refill
For Use With (Application) Purge and trap systems, to stop carrier gas interferences
For Use With (Equipment) Refillable Hydrocarbon trap
Includes Two recharges