Learn More
Restek™ Replacement O-Rings for High-Capacity Indicating Oxygen Trap
SDP
Catalog No. 06802903 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
O-ring
Catalog No. Type
06-802-903 O-ring
1 options
Catalog No. 06-802-903 Supplier Restek™ Supplier No. 22081
For high-capacity indicating oxygen trap.
  • Includes 5 large and 5 small o-rings
Specifications
Type O-ring
For Use With (Equipment) High capacity indicating Oxygen trap