Learn More
Restek™ Replacement Split/Splitless Injection Ports: Replacement Weldments
SDP
Catalog No. 06712017 Shop All Restek Corporation Products
Change view
Click to view available options
Product Type:
Shell Weldmen
Shell Weldment
Weldment
Weldment with EPC
Material:
Siltek Treated Stainless Steel
Stainless Steel
For Use With (Equipment):
Agilent 5890 GCs, Agilent 6890/6850 GCs
Agilent 6890/6850 GCs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Material For Use With (Equipment)
06-712-017 Weldment with EPC Siltek Treated Stainless Steel Agilent 6890/6850 GCs
06-710-552 Weldment Stainless Steel Agilent 5890 GCs, Agilent 6890/6850 GCs
06-712-018 Shell Weldmen Stainless Steel Agilent 6890/6850 GCs
06-712-016 Shell Weldment Siltek Treated Stainless Steel Agilent 6890/6850 GCs
06-712-019 Weldment with EPC Stainless Steel Agilent 6890/6850 GCs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 06-712-017 Supplier Restek™ Supplier No. 22672
Replacement weldments for Agilent 5890 and 6890/6850 GCs
  • Siltek™ treatment passivates metal surface ensuring an inert pathway for the sample, delivering increased performance
  • High-quality stainless steel injector
  • Meets or exceeds Agilent original equipment performance
Specifications
For Use With (Application) Gas Chromatography
For Use With (Equipment) Agilent 6890/6850 GCs
Material Siltek Treated Stainless Steel
Product Type Weldment with EPC