Learn More
Restek™ Replacement Split/Splitless Weldment
SDP
Catalog No. 06712028 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
06-712-028 Stainless Steel
1 options
Catalog No. 06-712-028 Supplier Restek™ Supplier No. 22686
Direct replacement for Agilent 5890 and 6890/6850 GCs
  • Siltek™ treatment passivates the metal surface ensuring an inert pathway for the sample, delivering increased performance
  • High-quality stainless steel injector
  • Meets or exceeds Agilent original equipment performance
Specifications
For Use With (Equipment) Large Canister Type Filter, Agilent 6890/6850 Gcs
Material Stainless Steel
Product Type Split/Splitless Weldment