Learn More
Restek™ Specialty Resprep™ EPA Method 548.1 Cartridge
SDP
Catalog No. 06713126 Shop All Restek Corporation Products
Encompass
Change view
Click to view available options
Volume (Metric):
6 mL
Catalog No. Volume (Metric)
06-713-126 6 mL
1 options
Catalog No. 06-713-126 Supplier Restek™ Supplier No. 26063
Extraction of endothal from aqueous samples
  • Weak anion exchange resin (BioRev 5)
Specifications
Certifications/Compliance EPA 548.1
For Use With (Application) Extraction of Endothal From Aqueous Samples
Type SPE Cartridge
Volume (Metric) 6 mL
Material HDPE, Polyethylene Frits
Resin Composition Weak AnIon Exchange (BioRev 5)