Learn More
Restek Corporation Restek Split Liners for Agilent/Finnigan GCs: Laminar Cup Splitter
Supplier: Restek Corporation 20801200.1
Sleeve, Laminar Cup Splitter; 4mm I.D.; For Agilent/Finnegan GC; w/FS Wool; Length: 78.5mm; 6.3mm O.D.