Learn More
Restek Corporation Restek Split Liners for Agilent/Finnigan GCs: mini-Lam Split
Supplier: Restek Corporation 20991200.5
Sleeve, mini-Lam Split; 4mm I.D.; For Agilent/Finnegan GC; w/FS Wool; Length: 78.5mm; 6.3mm O.D.; Pack of 5